4-Bromo-1H-indole-3-carbonitrile ≥95%

Supplier: Ambeed
Warning

A146845-250MG A146845-1G
77611-356EA 50.47 USD
77611-356 77683-254
4-Bromo-1H-indole-3-carbonitrile ≥95%
4-Bromo-1H-indole-3-carbonitrile

Precaución: For research use only.

Formula: C₉H₅BrN₂
MW: 221.05 g/mol
MDL Number: MFCD07787361
CAS Number: 903131-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR