2-(3-(4-Methylbenzoyl)phenyl)propanoic acid ≥90%

Supplier: Ambeed
Warning

Synonyms: Ketoprofen Related Compound A

A1466191-250MG A1466191-25MG
77875-034EA 624 USD
77875-034 77875-035
2-(3-(4-Methylbenzoyl)phenyl)propanoic acid ≥90%
2-(3-(4-Methylbenzoyl)phenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₇H₁₆O₃
MDL Number: MFCD27966872
CAS Number: 107257-20-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR