2,8-Diethyl-5,5-difluoro-1,3,7,9-tetramethyl-5H-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinin-4-ium-5-uide ≥98%

Supplier: Ambeed
Warning

Synonyms: {3-Ethyl-5-[(4-ethyl-3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]-2,4-dimethyl-1H-pyrrolato-N1,N5}difluoroboron

A1465633-100MG A1465633-1G A1465633-250MG
77597-358EA 45.1 USD
77597-358 77597-360 77597-362
2,8-Diethyl-5,5-difluoro-1,3,7,9-tetramethyl-5H-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinin-4-ium-5-uide ≥98%
2,8-Diethyl-5,5-difluoro-1,3,7,9-tetramethyl-5H-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinin-4-ium-5-uide

Precaución: for Research Use Only

Formula: C₁₇H₂₃BF₂N₂
MW: 304.1857 g/mol
MDL Number: MFCD28166503
CAS Number: 55799-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR