2-(((5-Bromothiophen-2-yl)methylene)amino)benzamide ≥97%

Supplier: Ambeed
Warning

A1464578-250MG A1464578-50MG A1464578-100MG
77875-012EA 426 USD
77875-012 77875-013 77875-011
2-(((5-Bromothiophen-2-yl)methylene)amino)benzamide ≥97%
2-(((5-Bromothiophen-2-yl)methylene)amino)benzamide

Precaución: for Research Use Only

Formula: C₁₂H₉BrN₂OS
MDL Number: MFCD00977636
CAS Number: 304456-62-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR