Hyaluronic acid sodium salt, M.W. ~1.5-1.8 x 10E6 Da

Supplier: Ambeed
Warning

Synonyms: Sodium hyaluronate, Hyaluronic acid sodium, HA sodium salt

A1463898-5G A1463898-25G A1463898-500G
77597-342EA 14.35 USD
77597-342 77624-224 77624-226
Hyaluronic acid sodium salt, M.W. ~1.5-1.8 x 10E6 Da
Hyaluronic acid sodium salt

Precaución: for Research Use Only

Formula: (C₁₄H₂₁NaNO₁₁)n
MDL Number: MFCD00875848
CAS Number: 9067-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR