3-(Cyclohex-1-en-1-yl)-6-(4-methoxyphenyl)-2-phenyl-5-(pyridin-2-ylamino)pyrazolo[1,5-a]pyrimidin-7(4H)-one ≥97%

Supplier: Ambeed
Warning

Synonyms: AG-270

A1462732-10MG A1462732-1MG A1462732-5MG
77597-318EA 146.58 USD
77597-318 77597-320 77597-322
3-(Cyclohex-1-en-1-yl)-6-(4-methoxyphenyl)-2-phenyl-5-(pyridin-2-ylamino)pyrazolo[1,5-a]pyrimidin-7(4H)-one ≥97%
3-(Cyclohex-1-en-1-yl)-6-(4-methoxyphenyl)-2-phenyl-5-(pyridin-2-ylamino)pyrazolo[1,5-a]pyrimidin-7(4H)-one

Precaución: for Research Use Only

Formula: C₃₀H₂₇N₅O₂
MW: 489.57 g/mol
MDL Number: MFCD34368529
CAS Number: 2201056-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR