rel-(1R-4E-pR)-Cyclooct-4-en-1-yl (4-nitrophenyl) carbonate ≥95%

Supplier: Ambeed
Warning

A1462408-100MG A1462408-250MG
77597-308EA 155.8 USD
77597-308 77597-310
rel-(1R-4E-pR)-Cyclooct-4-en-1-yl (4-nitrophenyl) carbonate ≥95%
rel-(1R-4E-pR)-Cyclooct-4-en-1-yl (4-nitrophenyl) carbonate

Precaución: for Research Use Only

Formula: C₁₅H₁₇NO₅
MW: 291.3 g/mol
CAS Number: 1354323-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR