Ethyl cellulose, 10 cP (50 mg/mL in ethanol:toluene = 1:4)

Supplier: Ambeed
Warning

Synonyms: Ethylcellulose

A1460272-100G A1460272-500G
77597-288EA 23.58 USD
77597-288 77597-290
Ethyl cellulose, 10 cP (50 mg/mL in ethanol:toluene = 1:4)
Ethyl cellulose

Precaución: for Research Use Only

MDL Number: MFCD00131037
CAS Number: 9004-57-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR