Aluminon AR

Supplier: Ambeed
Warning

Synonyms: ALuminon, Ammonium auritricarboxylate ATA Triammonium aurinetricarboxylate triammonium salt Ammonium aurintricarboxylate aurintricarboxylic acid ammonium salt Aluminon(C.1.No.43810) aurintricarboxylic acid triammonium salt, Ammonium 5,5'-((3-carboxylato-4-oxocyclohexa-2,5-dien-1-ylidene)methylene)bis(2-hydroxybenzoate), Aurin Tricarboxylic Acid Ammonium Salt

A1457765-100G A1457765-25G A1457765-500G
77597-278EA 52.28 USD
77597-278 77597-280 77622-188
Aluminon AR
Aluminon

Precaución: for Research Use Only

Formula: C₂₂H₂₃N₃O₉
MW: 473.44 g/mol
MDL Number: MFCD00040925
CAS Number: 569-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR