Polyethylene glycol methyl ether methacrylate, M.W. ~475 stabilised

Supplier: Ambeed
Warning

Synonyms: Polyethylene glycol monomethyl ether Methacrylate, mPEG Methacrylate, Methoxy PEG Methacrylate

Stabilization: (200:70-130) ppm Stabilised with BHT + MEHQ

A1456199-25G A1456199-500G
77874-939EA 19 USD
77874-939 77874-940
Polyethylene glycol methyl ether methacrylate, M.W. ~475 stabilised
Polyethylene glycol methyl ether methacrylate

Precaución: for Research Use Only

Formula: CH₃(OCH₂CH₂)nOCOC(=CH₂)CH₃
MDL Number: MFCD00241432
CAS Number: 26915-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR