Polyethylene glycol dimethacrylate Mn ~ 550 stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with BHT + MEHQ (270-330 : 80-120) ppm

A1456137-100G A1456137-25G
77874-937EA 20 USD
77874-937 77874-938
Polyethylene glycol dimethacrylate Mn ~ 550 stabilized
Polyethylene glycol dimethacrylate

Precaución: for Research Use Only

Formula: (C₂H₄O)nC₈H₁₀O₃
MDL Number: MFCD00014932
CAS Number: 25852-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR