Polyoxyethylene bis(amine), M.W. 4000
Supplier: Ambeed
|
Warning
|
Synonyms:
PEG-bis-amine, Amine-PEG-Amine, O,O′-Bis(2-aminoethyl)polyethylene glycol, PEG-diamine, Amine-PEG-amine (MW 4000), Poly(ethylene glycol) diamine, Polyoxyethylene bis(amine), Diaminopolyethylene glycol
A1456047-250MG
A1456047-100MG
A1456047-1G
77597-264EA
73.8
USD
77597-264
77597-260
77597-262
Polyoxyethylene bis(amine), M.W. 4000
Polyoxyethylene bis(amine)
Precaución: for Research Use Only
Formula:
H₂NCH₂CH₂(OCH₂CH₂)nNH₂ |
MDL Number:
MFCD00132004 CAS Number: 24991-53-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...