Cyanine 5 monoacid ≥95%

Supplier: Ambeed
Warning

Synonyms: Cy5 acid, 2-(5-(1-(5-Carboxypentyl)-3,3-dimethyl-5-sulphoindolin-2-ylidene)penta-1,3-dien-1-yl)-1-ethyl-3,3-dimethyl-3H-indol-1-ium-5-sulphonate

A1455193-100MG A1455193-1G
77597-258EA 287 USD
77597-258 77622-172
Cyanine 5 monoacid ≥95%
Cyanine 5 monoacid

Precaución: for Research Use Only

Formula: C₃₃H₄₀N₂O₈S₂
MW: 656.809 g/mol
MDL Number: MFCD29477740
CAS Number: 146368-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR