4-(Succinimidylcarboxy)benzophenone ≥98%

Supplier: Ambeed
Danger

Synonyms: 4-Benzoylbenzoic acid N-succinimidyl ester, 2,5-Dioxopyrrolidin-1-yl 4-benzoylbenzoate

A145504-100MG A145504-1G A145504-250MG
77748-015EA 43 USD
77748-015 77748-016 77748-017
4-(Succinimidylcarboxy)benzophenone ≥98%
4-(Succinimidylcarboxy)benzophenone

Precaución: for Research Use Only

Formula: C₁₈H₁₃NO₄
MDL Number: MFCD00058572
CAS Number: 91990-88-4
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR