5-Bromo-N-(4-chloro-3-(trifluoromethyl)phenyl)-1-(cyclopropanecarbonyl)indoline-6-sulfonamide ≥97%

Supplier: Ambeed
Warning

Synonyms: Indophagolin

A1453039-10MG A1453039-1MG A1453039-5MG
77597-236EA 26.65 USD
77597-236 77597-238 77597-240
5-Bromo-N-(4-chloro-3-(trifluoromethyl)phenyl)-1-(cyclopropanecarbonyl)indoline-6-sulfonamide ≥97%
5-Bromo-N-(4-chloro-3-(trifluoromethyl)phenyl)-1-(cyclopropanecarbonyl)indoline-6-sulfonamide

Precaución: for Research Use Only

Formula: C₁₉H₁₅BrClF₃N₂O₃S
MW: 523.75 g/mol
MDL Number: MFCD14721653
CAS Number: 1207660-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR