3-Bromo-5-chloro-2-methylthiophene ≥97%

Supplier: Ambeed
Warning

A1452997-25G A1452997-1G
77597-234EA 537.1 USD
77597-234 77874-920
3-Bromo-5-chloro-2-methylthiophene ≥97%
3-Bromo-5-chloro-2-methylthiophene

Precaución: for Research Use Only

Formula: C₅H₄BrClS
MDL Number: MFCD19687381
CAS Number: 61074-71-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR