(3R,4S,5S)-4-((S)-2-((S)-2-(Dimethylamino)-3-methylbutanamido)-N,3-dimethylbutanamido)-3-methoxy-5-methylheptanoic acid ≥97%

Supplier: Ambeed
Warning

A1452151-100MG A1452151-1G A1452151-250MG
77597-228EA 58.43 USD
77597-228 77597-230 77597-232
(3R,4S,5S)-4-((S)-2-((S)-2-(Dimethylamino)-3-methylbutanamido)-N,3-dimethylbutanamido)-3-methoxy-5-methylheptanoic acid ≥97%
(3R,4S,5S)-4-((S)-2-((S)-2-(Dimethylamino)-3-methylbutanamido)-N,3-dimethylbutanamido)-3-methoxy-5-methylheptanoic acid

Precaución: for Research Use Only

Formula: C₂₂H₄₃N₃O₅
MW: 429.59 g/mol
MDL Number: MFCD28387095
CAS Number: 133120-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR