LY-2835219 ≥98%

Supplier: Ambeed
Warning

Synonyms: N-(5-((4-Ethylpiperazin-1-yl)methyl)pyridin-2-yl)-5-fluoro-4-(4-fluoro-1-isopropyl-2-methyl-1H-benzo[d]imidazol-6-yl)pyrimidin-2-amine methanesulphonate, Abemaciclib, N-(5-((4-ethylpiperazin-1-yl)methyl)pyridin-2-yl)-5-fluoro-4-(4-fluoro-1-isopropyl-2-methyl-1H-benzo[d]imidazol-6-yl)pyrimidin-2-amine, Abemaciclib methanesulphonate

A144902-10MG A144902-250MG A144902-50MG A144902-1G
77611-332EA 6.18 USD
77611-332 77683-244 77683-245 77683-243
LY-2835219 ≥98%
LY-2835219

Precaución: For research use only.

Formula: C₂₇H₃₂F₂N₈·CH₄O₃S
MW: 602.7 g/mol
MDL Number: MFCD25562906
CAS Number: 1231930-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR