(R)-2,4-Diamino-4-oxobutanoic acid ≥97%

Supplier: Ambeed
Warning

Synonyms: (R)-2-Aminosuccinamic acid, H-D-Asn-OH, D-Asparagine

A1448497-1000G A1448497-500G A1448497-25G
77622-146EA 330.63 USD
77622-146 77622-148 77874-903
(R)-2,4-Diamino-4-oxobutanoic acid ≥97%
(R)-2,4-Diamino-4-oxobutanoic acid

Precaución: for Research Use Only

Formula: C₄H₈N₂O₃
MDL Number: MFCD00008036
CAS Number: 2058-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR