7-Fluoro-1H-indazole-4-boronic acid pinacol ester ≥98%

Supplier: Ambeed
Warning

Synonyms: 7-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole

A144790-100MG A144790-250MG
77685-229EA 80 USD
77685-229 77685-230
7-Fluoro-1H-indazole-4-boronic acid pinacol ester ≥98%
7-Fluoro-1H-indazole-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₆BFN₂O₂
MDL Number: MFCD19687061
CAS Number: 1186334-64-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR