1-Methylquinoxalin-2(1H)-one ≥98%

Supplier: Ambeed
Warning

A1445769-100MG A1445769-250MG
77622-138EA 17.51 USD
77622-138 77622-140
1-Methylquinoxalin-2(1H)-one ≥98%
1-Methylquinoxalin-2(1H)-one

Precaución: for Research Use Only

Formula: C₉H₈N₂O
MW: 160.1726 g/mol
MDL Number: MFCD01860250
CAS Number: 6479-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR