(2R,3R,4R,5R)-2-(Acetoxymethyl)-5-(2-oxo-4-thioxo-3,4-dihydropyrimidin-1(2H)-yl)tetrahydrofuran-3,4-diyl diacetate ≥98%

Supplier: Ambeed
Warning

A1445266-100MG A1445266-1G A1445266-250MG
77874-884EA 28 USD
77874-884 77874-885 77874-886
(2R,3R,4R,5R)-2-(Acetoxymethyl)-5-(2-oxo-4-thioxo-3,4-dihydropyrimidin-1(2H)-yl)tetrahydrofuran-3,4-diyl diacetate ≥98%
(2R,3R,4R,5R)-2-(Acetoxymethyl)-5-(2-oxo-4-thioxo-3,4-dihydropyrimidin-1(2H)-yl)tetrahydrofuran-3,4-diyl diacetate

Precaución: for Research Use Only

Formula: C₁₅H₁₈N₂O₈S
CAS Number: 55003-25-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR