1-(9H-Fluoren-9-yl)-3,6-dioxo-2,9-dioxa-4,7-diazaundecan-11-oic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Gly-NH-CH2-O-CH2COOH

A1443651-250MG A1443651-5G
77874-864EA 52 USD
77874-864 77874-865
1-(9H-Fluoren-9-yl)-3,6-dioxo-2,9-dioxa-4,7-diazaundecan-11-oic acid ≥98%
1-(9H-Fluoren-9-yl)-3,6-dioxo-2,9-dioxa-4,7-diazaundecan-11-oic acid

Precaución: for Research Use Only

Formula: C₂₀H₂₀N₂O₆
MDL Number: MFCD32639967
CAS Number: 1599440-08-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR