(2S,3R,4S,5S,6R)-2-(2,4-dihydroxy-6-((E)-4-hydroxystyryl)phenoxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol ≥98%

Supplier: Ambeed
Warning

Synonyms: 2,3,5,4'-Tetrahydroxystilbene 2-O-β-D-glucoside

A144286-1G A144286-250MG A144286-100MG
77611-324EA 138.02 USD
77611-324 77611-326 77685-225
(2S,3R,4S,5S,6R)-2-(2,4-dihydroxy-6-((E)-4-hydroxystyryl)phenoxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol ≥98%
(2S,3R,4S,5S,6R)-2-(2,4-dihydroxy-6-((E)-4-hydroxystyryl)phenoxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

Precaución: for Research Use Only

Formula: C₂₀H₂₂O₉
MW: 406.38 g/mol
MDL Number: MFCD00238694
CAS Number: 82373-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR