N-(2-((4-Chlorobenzyl)thio)benzo[d]thiazol-6-yl)-2-(morpholine-4-carbonyl)benzamide ≥97%

Supplier: Ambeed
Warning

Synonyms: CID1231538

A1441848-100MG A1441848-25MG
77622-088EA 183.34 USD
77622-088 77622-090
N-(2-((4-Chlorobenzyl)thio)benzo[d]thiazol-6-yl)-2-(morpholine-4-carbonyl)benzamide ≥97%
N-(2-((4-Chlorobenzyl)thio)benzo[d]thiazol-6-yl)-2-(morpholine-4-carbonyl)benzamide

Precaución: for Research Use Only

Formula: C₂₆H₂₂ClN₃O₃S₂
MW: 524.0542 g/mol
MDL Number: MFCD02071667
CAS Number: 354126-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR