(Z)-N'-Hydroxybenzimidamide hydrobromide ≥98%

Supplier: Ambeed
Warning

A1441266-1G A1441266-100MG A1441266-250MG
77624-198EA 41.2 USD
77624-198 77874-830 77874-831
(Z)-N'-Hydroxybenzimidamide hydrobromide ≥98%
(Z)-N'-Hydroxybenzimidamide hydrobromide

Precaución: for Research Use Only

Formula: C₇H₉BrN₂O
MDL Number: MFCD00031485
CAS Number: 1195196-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR