(R)-tert-Butyl (3-amino-2-hydroxypropyl)carbamate ≥90%

Supplier: Ambeed
Danger

A1440324-250MG A1440324-5G
77622-072EA 286.34 USD
77622-072 77874-823
(R)-tert-Butyl (3-amino-2-hydroxypropyl)carbamate ≥90%
(R)-tert-Butyl (3-amino-2-hydroxypropyl)carbamate

Precaución: for Research Use Only

Formula: C₈H₁₈N₂O₃
MDL Number: MFCD32736306
CAS Number: 1042665-73-5
UN: 3263
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR