2,6-Dichloro-N-(3-isobutyl-1,5-dimethyl-1H-pyrazol-4-yl)-4-(2-(piperazin-1-yl)pyridin-4-yl)benzenesulfonamide ≥97%

Supplier: Ambeed
Warning

Synonyms: PCLX-001

A1440111-10MG A1440111-25MG A1440111-50MG
77874-818EA 836 USD
77874-818 77874-819 77874-820
2,6-Dichloro-N-(3-isobutyl-1,5-dimethyl-1H-pyrazol-4-yl)-4-(2-(piperazin-1-yl)pyridin-4-yl)benzenesulfonamide ≥97%
2,6-Dichloro-N-(3-isobutyl-1,5-dimethyl-1H-pyrazol-4-yl)-4-(2-(piperazin-1-yl)pyridin-4-yl)benzenesulfonamide

Precaución: for Research Use Only

Formula: C₂₄H₃₀Cl₂N₆O₂S
CAS Number: 1215011-08-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR