2,2'-(Cyclopropane-1,1-diyl)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane) ≥97%

Supplier: Ambeed
Warning

A1439617-100MG A1439617-250MG
77874-808EA 55 USD
77874-808 77874-809
2,2'-(Cyclopropane-1,1-diyl)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane) ≥97%
2,2'-(Cyclopropane-1,1-diyl)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane)

Precaución: for Research Use Only

Formula: C₁₅H₂₈B₂O₄
MDL Number: MFCD32648877
CAS Number: 2222867-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR