(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-methyl-1H-indol-3-yl)propanoic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Trp(4-Me)-OH

A1438047-1G A1438047-250MG A1438047-5G
77874-798EA 381 USD
77874-798 77874-799 77874-800
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-methyl-1H-indol-3-yl)propanoic acid ≥98%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-methyl-1H-indol-3-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₂₇H₂₄N₂O₄
MW: 440.49 g/mol
MDL Number: MFCD01632012
CAS Number: 2411871-30-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR