(4-(Benzyloxy)-3-(trifluoromethyl)phenyl)boronic acid ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: 4-(Benzyloxy)-3-(trifluoromethyl)phenylboronic acid

A143306-1G A143306-100MG A143306-250MG A143306-5G
77683-237EA 45 USD
77683-237 77885-204 77885-205 77885-206
(4-(Benzyloxy)-3-(trifluoromethyl)phenyl)boronic acid ≥95%, Ambeed.Inc
(4-(Benzyloxy)-3-(trifluoromethyl)phenyl)boronic acid

Precaución: For research use only.

Formula: C₁₄H₁₂BF₃O₃
MDL Number: MFCD18383520
CAS Number: 1245014-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR