4-Iodo-3,5-dimethylbenzonitrile ≥95%

Supplier: Ambeed
Warning

A143295-100MG A143295-1G A143295-250MG
77885-201EA 10 USD
77885-201 77885-202 77885-203
4-Iodo-3,5-dimethylbenzonitrile ≥95%
4-Iodo-3,5-dimethylbenzonitrile

Precaución: for Research Use Only

Formula: C₉H₈IN
MDL Number: MFCD19441144
CAS Number: 1227311-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR