1-Bromo-4-methylisoquinoline ≥97%, Ambeed.Inc

Supplier: Ambeed
Warning

A143030-1G A143030-250MG
77683-236EA 383 USD
77683-236 77889-954
1-Bromo-4-methylisoquinoline ≥97%, Ambeed.Inc
1-Bromo-4-methylisoquinoline

Precaución: For research use only.

Formula: C₁₀H₈BrN
MDL Number: MFCD22096517
CAS Number: 1542564-70-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR