Methyl 1H-pyrrolo[2,3-c]pyridine-3-carboxylate ≥98%

Supplier: Ambeed
Warning

A142555-1G A142555-250MG A142555-5G
77611-298EA 67.4 USD
77611-298 77883-146 77883-147
Methyl 1H-pyrrolo[2,3-c]pyridine-3-carboxylate ≥98%
Methyl 1H-pyrrolo[2,3-c]pyridine-3-carboxylate

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MDL Number: MFCD16036494
CAS Number: 108128-12-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR