Ampicillin trihydrate ≥99%

Supplier: Ambeed
Danger

A142546-100G A142546-5G A142546-25G
77600-064EA 58.43 USD
77600-064 77624-188 77876-980
Ampicillin trihydrate ≥99%
Ampicillin trihydrate

Precaución: for Research Use Only

Formula: C₁₆H₁₉N₃O₄S·3H₂O
MDL Number: MFCD00072036
CAS Number: 7177-48-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR