6-Bromo-5-methoxy-1H-indole ≥95%

Supplier: Ambeed
Warning

A142341-10G A142341-250MG
77883-143EA 1055 USD
77883-143 77883-144
6-Bromo-5-methoxy-1H-indole ≥95%
6-Bromo-5-methoxy-1H-indole

Precaución: for Research Use Only

Formula: C₉H₈BrNO
MDL Number: MFCD11848542
CAS Number: 106103-36-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR