(4S,5'R,6aR,6bS,8aS,8bR,9S,10R,11aS,12aS,12bS)-5',6a,8a,9-Tetramethyl-1,3,4,5,6,6a,6b,7,8,8a,8b,9,11a,12,12a,12b-hexadecahydrospiro[naphtho[2',1':4,5]indeno[2,1-b]furan-10,2'-piperidin]-4-ol ≥98%

Supplier: Ambeed
Warning

Synonyms: Solasodine

A142284-10MG A142284-1MG
77592-688EA 44.08 USD
77592-688 77600-062
(4S,5'R,6aR,6bS,8aS,8bR,9S,10R,11aS,12aS,12bS)-5',6a,8a,9-Tetramethyl-1,3,4,5,6,6a,6b,7,8,8a,8b,9,11a,12,12a,12b-hexadecahydrospiro[naphtho[2',1':4,5]indeno[2,1-b]furan-10,2'-piperidin]-4-ol ≥98%
(4S,5'R,6aR,6bS,8aS,8bR,9S,10R,11aS,12aS,12bS)-5',6a,8a,9-Tetramethyl-1,3,4,5,6,6a,6b,7,8,8a,8b,9,11a,12,12a,12b-hexadecahydrospiro[naphtho[2',1':4,5]indeno[2,1-b]furan-10,2'-piperidin]-4-ol

Precaución: for Research Use Only

Formula: C₂₇H₄₃NO₂
MW: 413.64 g/mol
MDL Number: MFCD00037844
CAS Number: 126-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR