4-((4-(2,4-Dichlorophenyl)-3-ethylthiazol-2(3H)-ylidene)amino)benzenesulfonamide ≥97%

Supplier: Ambeed
Warning

Synonyms: EMAC10101d

A1417612-10MG A1417612-1MG A1417612-5MG
77597-170EA 50.23 USD
77597-170 77597-172 77597-174
4-((4-(2,4-Dichlorophenyl)-3-ethylthiazol-2(3H)-ylidene)amino)benzenesulfonamide ≥97%
4-((4-(2,4-Dichlorophenyl)-3-ethylthiazol-2(3H)-ylidene)amino)benzenesulfonamide

Precaución: for Research Use Only

Formula: C₁₇H₁₅Cl₂N₃O₂S₂
MW: 428.36 g/mol
MDL Number: MFCD34165711
CAS Number: 2561476-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR