5-Benzyl-5-azaspiro[2.4]heptane-4,7-dione ≥98%

Supplier: Ambeed
Warning

A141469-1G A141469-250MG
77615-126EA 51.5 USD
77615-126 77746-000
5-Benzyl-5-azaspiro[2.4]heptane-4,7-dione ≥98%
5-Benzyl-5-azaspiro[2.4]heptane-4,7-dione

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO₂
MW: 215.24 g/mol
MDL Number: MFCD22050713
CAS Number: 129306-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR