1-Methylpiperidine-4-boronic acid 95+%

Supplier: Ambeed
Warning

A141356-1G A141356-250MG A141356-50MG
77885-185EA 567 USD
77885-185 77885-186 77885-187
1-Methylpiperidine-4-boronic acid 95+%
1-Methylpiperidine-4-boronic acid

Precaución: for Research Use Only

Formula: C₆H₁₄BNO₂
MDL Number: MFCD07371554
CAS Number: 706748-53-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR