(1R,2R)-2-Amino-1-methylcyclopentan-1-ol ≥98%

Supplier: Ambeed
Danger

A1412506-250MG A1412506-100MG
77592-682EA 94.3 USD
77592-682 77592-680
(1R,2R)-2-Amino-1-methylcyclopentan-1-ol ≥98%
(1R,2R)-2-Amino-1-methylcyclopentan-1-ol

Precaución: for Research Use Only

Formula: C₆H₁₃NO
MW: 115.17 g/mol
MDL Number: MFCD19205405
CAS Number: 1400689-45-3
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR