1-Benzyl-N5-cyclopropyl-N3-methyl-2-oxo-1,2-dihydropyridine-3,5-dicarboxamide ≥97%

Supplier: Ambeed
Warning

Synonyms: GSK620

A1411591-10MG A1411591-1MG A1411591-5MG
77597-158EA 66.63 USD
77597-158 77597-160 77597-162
1-Benzyl-N5-cyclopropyl-N3-methyl-2-oxo-1,2-dihydropyridine-3,5-dicarboxamide ≥97%
1-Benzyl-N5-cyclopropyl-N3-methyl-2-oxo-1,2-dihydropyridine-3,5-dicarboxamide

Precaución: for Research Use Only

Formula: C₁₈H₁₉N₃O₃
CAS Number: 2088410-46-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR