2-Chloro-6-fluorobenzo[d]oxazole ≥95%

Supplier: Ambeed
Warning

A141134-1G A141134-250MG
77883-129EA 63 USD
77883-129 77883-130
2-Chloro-6-fluorobenzo[d]oxazole ≥95%
2-Chloro-6-fluorobenzo[d]oxazole

Precaución: for Research Use Only

Formula: C₇H₃ClFNO
MDL Number: MFCD16250625
CAS Number: 153403-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR