2-Fluorocyclobutan-1-amine hydrochloride ≥97%
Supplier: Ambeed
|
Warning
|
A1410209-100MG
A1410209-1G
A1410209-5G
77874-768EA
131
USD
77874-768
77874-769
77874-770
2-Fluorocyclobutan-1-amine hydrochloride ≥97%
2-Fluorocyclobutan-1-amine hydrochloride
Precaución: for Research Use Only
Formula:
C₄H₉ClFN |
MDL Number:
MFCD31743178 CAS Number: 2288709-22-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...