2-(((3aR,4S,6R,6aS)-6-(7-(((1R,2S)-2-(3,4-Difluorophenyl)cyclopropyl)amino)-5-(propylthio)-3H-[1,2,3]triazolo[4,5-d]pyrimidin-3-yl)-2,2-dimethyltetrahydro-3aH-cyclopenta[d][1,3]dioxol-4-yl)oxy)ethanol ≥95%

Supplier: Ambeed
Warning

A140979-10G A140979-250MG A140979-5G
77891-983EA 295 USD
77891-983 77891-984 77891-985
2-(((3aR,4S,6R,6aS)-6-(7-(((1R,2S)-2-(3,4-Difluorophenyl)cyclopropyl)amino)-5-(propylthio)-3H-[1,2,3]triazolo[4,5-d]pyrimidin-3-yl)-2,2-dimethyltetrahydro-3aH-cyclopenta[d][1,3]dioxol-4-yl)oxy)ethanol ≥95%
2-(((3aR,4S,6R,6aS)-6-(7-(((1R,2S)-2-(3,4-Difluorophenyl)cyclopropyl)amino)-5-(propylthio)-3H-[1,2,3]triazolo[4,5-d]pyrimidin-3-yl)-2,2-dimethyltetrahydro-3aH-cyclopenta[d][1,3]dioxol-4-yl)oxy)ethanol

Precaución: for Research Use Only

Formula: C₂₆H₃₂F₂N₆O₄S
MW: 562.63 g/mol
MDL Number: MFCD23160137
CAS Number: 274693-26-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR