tert-Butyl 4-iodobutanoate ≥98%

Supplier: Ambeed
Warning

A1409637-1G A1409637-5G A1409637-250MG A1409637-100MG
77597-154EA 230.63 USD
77597-154 77597-156 77622-048 77874-763
tert-Butyl 4-iodobutanoate ≥98%

Precaución: for Research Use Only

Formula: C₈H₁₅IO₂
MDL Number: MFCD25965404
CAS Number: 6182-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR