3-Methoxycyclobutan-1-ol ≥95%

Supplier: Ambeed
Warning

A140760-100MG A140760-250MG
77747-990EA 73 USD
77747-990 77747-991
3-Methoxycyclobutan-1-ol ≥95%
3-Methoxycyclobutan-1-ol

Precaución: for Research Use Only

Formula: C₅H₁₀O₂
MDL Number: MFCD21756147
CAS Number: 1432680-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR