Laurdan ≥98%

Supplier: Ambeed
Warning

A140750-100MG A140750-250MG A140750-50MG
77667-030EA 282 USD
77667-030 77878-992 77878-993
Laurdan ≥98%
Laurdan

Precaución: for Research Use Only

Formula: C₂₄H₃₅NO
MDL Number: MFCD00467560
CAS Number: 74515-25-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR