(S)-1-(tert-Butoxycarbonyl)-4,4-dimethylpyrrolidine-2-carboxylic acid ≥98%

Supplier: Ambeed
Warning

A140716-100MG A140716-250MG
77901-681EA 56 USD
77901-681 77901-406
(S)-1-(tert-Butoxycarbonyl)-4,4-dimethylpyrrolidine-2-carboxylic acid ≥98%
(S)-1-(tert-Butoxycarbonyl)-4,4-dimethylpyrrolidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₄
MDL Number: MFCD11040193
CAS Number: 1001353-87-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR