[-]-Deguelin ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A139886-1MG A139886-10MG A139886-5MG A139886-100MG A139886-50MG
77611-274EA 16.48 USD
77611-274 77683-220 77683-221 77891-967 77891-968
[-]-Deguelin ≥98%, Ambeed.Inc
[-]-Deguelin

Precaución: For research use only.

Formula: C₂₃H₂₂O₆
MDL Number: MFCD01740600
CAS Number: 522-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR